The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320728.1 | NP_001307657.1 | 1 - 2 | 209 | 130 |
NM_024610.5 | NP_078886.2 | 1 - 2 | 201 | 130 | |
XM_005247762.2 | XP_005247819.1 | 1 - 2 | 201 | 130 | |
GenBank mRNA | AF400663.1 | - | 1 - 2 | 168 | 130 |
AK289410.1 | - | 1 - 2 | 201 | 130 | |
BC011897.2 | - | 1 - 2 | 185 | 130 | |
BC063629.1 | - | 1 - 2 | 141 | 130 | |
BI912690.1 | - | 1 - 2 | 181 | 130 | |
HY020808.1 | - | 1 - 2 | 130 |