Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAD51L3-RFFL
Entrez Gene ID
100529207
Gene Name
RAD51L3-RFFL readthrough
Gene Aliases
-
Chromosome Location
Chr.17: 35009112 - 35121522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037714.1 5 - 6 650 124
GenBank mRNA AK097811.1 - 5 - 6 621 124

View Full Product Details