The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289003.1 | NP_001275932.1 | 8 - 9 | 1994 | 107 |
NM_015020.3 | NP_055835.2 | 8 - 9 | 1647 | 107 | |
GenBank mRNA | AB023148.2 | - | 8 - 9 | 1994 | 107 |
BC035267.1 | - | 3 - 4 | 482 | 107 | |
BC129927.1 | - | 7 - 8 | 1274 | 107 | |
BX647823.1 | - | 8 - 9 | 1648 | 107 | |
BX648157.1 | - | 2 - 3 | 384 | 107 |