The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000888.4 | NP_000879.2 | 13 - 14 | 2353 | 77 |
NM_001282353.1 | NP_001269282.1 | 14 - 15 | 2246 | 77 | |
NM_001282354.1 | NP_001269283.1 | 11 - 12 | 2068 | 77 | |
NM_001282355.1 | NP_001269284.1 | 11 - 12 | 2032 | 77 | |
NM_001282388.1 | NP_001269317.1 | 12 - 13 | 2273 | 77 | |
NM_001282389.1 | NP_001269318.1 | 13 - 14 | 2180 | 77 | |
NM_001282390.1 | NP_001269319.1 | 12 - 13 | 2100 | 77 | |
GenBank mRNA | AK290300.1 | - | 13 - 14 | 2321 | 77 |
AK313944.1 | - | 14 - 15 | 2237 | 77 | |
AL831998.1 | - | 13 - 14 | 2292 | 77 | |
BC121178.2 | - | 13 - 14 | 2123 | 77 | |
BC143656.1 | - | 10 - 11 | 1488 | 77 | |
CD013944.1 | - | 12 - 13 | 2045 | 77 | |
CD013945.1 | - | 11 - 12 | 1977 | 77 | |
CD013946.1 | - | 11 - 12 | 2013 | 77 | |
CD013947.1 | - | 13 - 14 | 2125 | 77 | |
M35198.3 | - | 13 - 14 | 2119 | 77 |