The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284240.1 | NP_001271169.1 | 3 - 4 | 1873 | 103 |
NM_018999.3 | NP_061872.2 | 3 - 4 | 1873 | 103 | |
XM_005269905.3 | XP_005269962.1 | 3 - 4 | 1904 | 103 | |
XM_005269906.3 | XP_005269963.1 | 3 - 4 | 1904 | 103 | |
XM_011539870.2 | XP_011538172.1 | 3 - 4 | 1904 | 103 | |
XM_011539871.2 | XP_011538173.1 | 3 - 4 | 1904 | 103 | |
XM_011539872.2 | XP_011538174.1 | 3 - 4 | 1889 | 103 | |
XM_017016340.1 | XP_016871829.1 | 3 - 4 | 1793 | 103 | |
XM_017016341.1 | XP_016871830.1 | 3 - 4 | 1889 | 103 | |
GenBank mRNA | AB032954.1 | - | 2 - 3 | 876 | 103 |
AF241785.1 | - | 2 - 3 | 1362 | 103 | |
AK024324.1 | - | 2 - 3 | 1597 | 103 | |
CR991891.1 | - | 2 - 3 | 526 | 103 |