The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020666.2 | NP_065717.1 | 6 - 7 | 766 | 110 |
GenBank mRNA | AF294429.1 | - | 5 - 6 | 658 | 110 |
AK311485.1 | - | 5 - 6 | 677 | 110 | |
BC015942.1 | - | 6 - 7 | 766 | 110 | |
BC136261.1 | - | 6 - 7 | 714 | 110 | |
BC143547.1 | - | 6 - 7 | 714 | 110 | |
BC151233.1 | - | 6 - 7 | 1980 | 110 | |
BX647323.1 | - | 2 - 3 | 623 | 110 | |
CR749504.1 | - | 8 - 9 | 1645 | 110 |