The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301007.1 | NP_001287936.1 | 9 - 10 | 2271 | 95 |
NM_130847.2 | NP_570899.1 | 10 - 11 | 2421 | 95 | |
XM_005273798.4 | XP_005273855.1 | 12 - 13 | 2564 | 95 | |
XM_005273801.4 | XP_005273858.1 | 10 - 11 | 2257 | 95 | |
XM_011542626.2 | XP_011540928.1 | 11 - 12 | 2414 | 95 | |
XM_011542628.2 | XP_011540930.1 | 8 - 9 | 1725 | 95 | |
GenBank mRNA | AK074084.1 | - | 10 - 11 | 2389 | 95 |
AL833833.1 | - | 3 - 4 | 362 | 95 | |
BC014126.1 | - | 7 - 8 | 878 | 95 | |
BC037539.1 | - | 10 - 11 | 2392 | 95 | |
BC040683.1 | - | 1 - 2 | 115 | 95 | |
BX648729.1 | - | 9 - 10 | 2271 | 95 |