Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC29A3
Entrez Gene ID
55315
Gene Name
solute carrier family 29 member 3
Gene Aliases
ENT3, HCLAP, HJCD, PHID
Chromosome Location
Chr.10: 71319253 - 71363390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174098.1 NP_001167569.1 3 - 4 443 72
NM_018344.5 NP_060814.4 3 - 4 443 72
XM_006717910.3 XP_006717973.1 3 - 4 835 72
GenBank mRNA AF326987.1 - 2 - 3 391 72
AK002022.1 - 2 - 3 194 72
AK314497.1 - 3 - 4 411 72
AX775915.1 - 3 - 4 443 72
AX775917.1 - 3 - 4 443 72
AX775919.1 - 3 - 4 443 72
AX775921.1 - 3 - 4 443 72
AY288928.1 - 3 - 4 403 72
AY358686.1 - 3 - 4 421 72
BC041575.1 - 2 - 3 331 72
BC120996.1 - 3 - 4 421 72
BC120997.1 - 3 - 4 421 72
DC319920.1 - 3 - 4 443 72

View Full Product Details