The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308171.1 | NP_001295100.1 | 8 - 9 | 810 | 115 |
NM_005925.2 | NP_005916.2 | 8 - 9 | 810 | 115 | |
GenBank mRNA | AY695931.1 | - | 8 - 9 | 821 | 115 |
BC136559.1 | - | 8 - 9 | 788 | 115 | |
BC144244.1 | - | 8 - 9 | 788 | 115 | |
X81333.1 | - | 8 - 9 | 809 | 115 |