The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147156.3 | NP_671512.1 | 10 - 11 | 2016 | 67 |
XM_005269675.1 | XP_005269732.1 | 5 - 6 | 1708 | 67 | |
XM_011539582.2 | XP_011537884.1 | 7 - 8 | 1620 | 67 | |
XM_011539583.2 | XP_011537885.1 | 5 - 6 | 1497 | 67 | |
GenBank mRNA | AB154421.1 | - | 6 - 7 | 1399 | 67 |
AK026683.1 | - | 6 - 7 | 1479 | 67 | |
AK296233.1 | - | 4 - 5 | 683 | 67 | |
AY280959.1 | - | 10 - 11 | 1982 | 67 | |
AY312431.1 | - | 4 - 5 | 1208 | 67 | |
BC042899.1 | - | 4 - 5 | 1277 | 67 |