The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147156.3 | NP_671512.1 | 4 - 5 | 504 | 152 |
GenBank mRNA | AK311737.1 | - | 4 - 5 | 520 | 152 |
AY280959.1 | - | 4 - 5 | 470 | 152 | |
AY332650.1 | - | 4 - 5 | 379 | 152 | |
AY364088.1 | - | 4 - 5 | 379 | 152 | |
BM764008.1 | - | 4 - 5 | 503 | 152 | |
KP710931.1 | - | 3 - 4 | 193 | 152 | |
KP710932.1 | - | 3 - 4 | 193 | 152 | |
KR020014.1 | - | 2 - 3 | 184 | 152 | |
KR020022.1 | - | 4 - 5 | 420 | 152 |