The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166139.2 | NP_001159611.1 | 5 - 6 | 805 | 136 |
XM_017007964.1 | XP_016863453.1 | 6 - 7 | 681 | 136 |