The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003462.3 | NP_003453.2 | 4 - 5 | 655 | 102 |
XM_005271172.2 | XP_005271229.1 | 3 - 4 | 467 | 102 | |
GenBank mRNA | AF006386.1 | - | 4 - 5 | 632 | 102 |
AI128097.1 | - | 2 - 3 | 102 | ||
AK225935.1 | - | 3 - 4 | 2694 | 102 | |
AK290502.1 | - | 4 - 5 | 579 | 102 | |
AK295199.1 | - | 3 - 4 | 398 | 102 | |
BC039074.1 | - | 4 - 5 | 638 | 102 | |
BC046117.1 | - | 4 - 5 | 605 | 102 | |
BX647913.1 | - | 4 - 5 | 655 | 102 |