Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORL1
Entrez Gene ID
6653
Gene Name
sortilin related receptor 1
Gene Aliases
C11orf32, LR11, LRP9, SORLA, SorLA-1, gp250
Chromosome Location
Chr.11: 121452203 - 121633762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003105.5 NP_003096.1 16 - 17 2397 84
XM_011542963.2 XP_011541265.1 16 - 17 2272 84
XM_011542965.2 XP_011541267.1 10 - 11 1306 84
XM_017018169.1 XP_016873658.1 16 - 17 2104 84
XM_017018170.1 XP_016873659.1 14 - 15 1780 84
XM_017018171.1 XP_016873660.1 16 - 17 2272 84
XM_017018172.1 XP_016873661.1 3 - 4 361 84
GenBank mRNA BC137171.1 - 16 - 17 2320 84
Y08110.1 - 16 - 17 2348 84

View Full Product Details