The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003105.5 | NP_003096.1 | 44 - 45 | 6189 | 112 |
XM_011542963.2 | XP_011541265.1 | 43 - 44 | 5950 | 112 | |
XM_011542965.2 | XP_011541267.1 | 38 - 39 | 5098 | 112 | |
XM_011542967.2 | XP_011541269.1 | 24 - 25 | 3130 | 112 | |
XM_017018169.1 | XP_016873658.1 | 44 - 45 | 5896 | 112 | |
XM_017018170.1 | XP_016873659.1 | 42 - 43 | 5572 | 112 | |
XM_017018172.1 | XP_016873661.1 | 30 - 31 | 4061 | 112 | |
GenBank mRNA | AK294404.1 | - | 15 - 16 | 2327 | 112 |
AK295878.1 | - | 24 - 25 | 3130 | 112 | |
AK302589.1 | - | 21 - 22 | 2801 | 112 | |
AK316340.1 | - | 15 - 16 | 1909 | 112 | |
BC137171.1 | - | 44 - 45 | 6112 | 112 | |
Y08110.1 | - | 44 - 45 | 6140 | 112 |