Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JUP
Entrez Gene ID
3728
Gene Name
junction plakoglobin
Gene Aliases
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB
Chromosome Location
Chr.17: 41754607 - 41786853 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002230.2 NP_002221.1 10 - 11 1903 59
NM_021991.2 NP_068831.1 10 - 11 1903 59
XM_006721873.2 XP_006721936.1 11 - 12 2153 59
XM_006721874.2 XP_006721937.1 10 - 11 2066 59
XM_006721875.1 XP_006721938.1 10 - 11 1985 59
XM_006721878.1 XP_006721941.1 10 - 11 1966 59
XM_011524753.2 XP_011523055.1 11 - 12 2189 59
XM_011524755.1 XP_011523057.1 10 - 11 1898 59
XM_011524756.1 XP_011523058.1 10 - 11 1796 59
XM_011524757.2 XP_011523059.1 10 - 11 2063 59
XM_011524758.1 XP_011523060.1 10 - 11 1792 59
XM_017024588.1 XP_016880077.1 10 - 11 1885 59
XM_017024589.1 XP_016880078.1 10 - 11 1886 59
XM_017024590.1 XP_016880079.1 10 - 11 1792 59
GenBank mRNA AK226051.1 - 10 - 11 1880 59
AY243535.1 - 9 - 10 1776 59
BC000441.2 - 10 - 11 1889 59
BC011865.2 - 10 - 11 2047 59
KU178155.1 - 9 - 10 1770 59
KU178156.1 - 4 - 5 537 59
M23410.1 - 10 - 11 1886 59
Z68228.1 - 10 - 11 1889 59

View Full Product Details