Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SCMH1
- Entrez Gene ID
-
22955
- Gene Name
- sex comb on midleg homolog 1 (Drosophila)
- Gene Aliases
-
Scml3
- Chromosome Location
-
Chr.1: 41027199 - 41242155 on Build GRCh38
View Full Product Details