Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMOD1
Entrez Gene ID
7111
Gene Name
tropomodulin 1
Gene Aliases
D9S57E, ETMOD, TMOD
Chromosome Location
Chr.9: 97501180 - 97601743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166116.1 NP_001159588.1 7 - 8 857 110
NM_003275.3 NP_003266.1 7 - 8 934 110
GenBank mRNA AK095748.1 - 4 - 5 601 110
AK314533.1 - 7 - 8 857 110
BC002660.2 - 7 - 8 934 110
M77016.1 - 6 - 7 755 110

View Full Product Details