Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRPSAP2
Entrez Gene ID
5636
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 2
Gene Aliases
PAP41
Chromosome Location
Chr.17: 18856299 - 18931287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243936.1 NP_001230865.1 8 - 9 949 71
NM_001243941.1 NP_001230870.1 8 - 9 983 71
NM_001243942.1 NP_001230871.1 9 - 10 1016 71
NM_002767.3 NP_002758.1 10 - 11 1132 71
XM_005256725.2 XP_005256782.1 9 - 10 986 71
XM_005256726.1 XP_005256783.1 9 - 10 1020 71
XM_005256729.4 XP_005256786.1 8 - 9 882 71
XM_017024864.1 XP_016880353.1 10 - 11 996 71
XM_017024865.1 XP_016880354.1 11 - 12 1205 71
XM_017024866.1 XP_016880355.1 11 - 12 1139 71
XM_017024867.1 XP_016880356.1 9 - 10 973 71
XM_017024868.1 XP_016880357.1 10 - 11 1850 71
XM_017024869.1 XP_016880358.1 10 - 11 1129 71
XM_017024870.1 XP_016880359.1 10 - 11 1044 71
XM_017024871.1 XP_016880360.1 9 - 10 1029 71
XM_017024872.1 XP_016880361.1 10 - 11 1165 71
XM_017024873.1 XP_016880362.1 9 - 10 1014 71
XM_017024874.1 XP_016880363.1 9 - 10 1086 71
XM_017024875.1 XP_016880364.1 9 - 10 959 71
XM_017024876.1 XP_016880365.1 8 - 9 937 71
GenBank mRNA AB007851.1 - 9 - 10 897 71
AK303911.1 - 8 - 9 895 71
AK304544.1 - 8 - 9 868 71
AK313087.1 - 10 - 11 1069 71
BC101670.1 - 9 - 10 858 71
BC101672.1 - 9 - 10 858 71
BC106050.1 - 10 - 11 1048 71
BX648850.1 - 9 - 10 965 71
CR457082.1 - 8 - 9 810 71

View Full Product Details