Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PRPSAP2
- Entrez Gene ID
-
5636
- Gene Name
- phosphoribosyl pyrophosphate synthetase associated protein 2
- Gene Aliases
-
PAP41
- Chromosome Location
-
Chr.17: 18856299 - 18931287 on Build GRCh38
View Full Product Details