The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013432.4 | NP_038460.4 | 12 - 13 | 1577 | 110 |
XM_011517048.2 | XP_011515350.1 | 5 - 6 | 676 | 110 | |
XM_011517049.2 | XP_011515351.1 | 5 - 6 | 637 | 110 | |
XM_011517050.2 | XP_011515352.1 | 12 - 13 | 1636 | 110 | |
GenBank mRNA | AB209126.1 | - | 4 - 5 | 2537 | 110 |
BC008782.2 | - | 11 - 12 | 1549 | 110 |