The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013432.4 | NP_038460.4 | 20 - 21 | 3103 | 143 |
XM_011517048.2 | XP_011515350.1 | 13 - 14 | 2202 | 143 | |
XM_011517049.2 | XP_011515351.1 | 13 - 14 | 2163 | 143 | |
GenBank mRNA | AB209126.1 | - | 12 - 13 | 4063 | 143 |
BC008782.2 | - | 19 - 20 | 3075 | 143 |