The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013432.4 | NP_038460.4 | 21 - 22 | 3403 | 149 |
XM_011517048.2 | XP_011515350.1 | 14 - 15 | 2502 | 149 | |
XM_011517049.2 | XP_011515351.1 | 14 - 15 | 2463 | 149 | |
GenBank mRNA | AB209126.1 | - | 13 - 14 | 4363 | 149 |
AK097406.1 | - | 1 - 2 | 291 | 149 | |
BC008782.2 | - | 20 - 21 | 3375 | 149 |