The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018982.4 | NP_061855.1 | 10 - 11 | 1307 | 62 |
NR_135075.1 | 8 - 9 | 1024 | 62 | ||
XM_017001499.1 | XP_016856988.1 | 9 - 10 | 1065 | 62 | |
XM_017001500.1 | XP_016856989.1 | 9 - 10 | 1128 | 62 | |
XM_017001501.1 | XP_016856990.1 | 8 - 9 | 909 | 62 | |
GenBank mRNA | AK000823.1 | - | 8 - 9 | 972 | 62 |
BC009674.1 | - | 5 - 6 | 620 | 62 | |
BC064590.1 | - | 10 - 11 | 1273 | 62 |