The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031694.2 | NP_001026864.1 | 3 - 4 | 302 | 139 |
NR_135092.1 | 3 - 4 | 302 | 139 | ||
GenBank mRNA | AK301497.1 | - | 1 - 2 | 144 | 139 |
BC009752.2 | - | 2 - 3 | 244 | 139 |