Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAIAP2L1
Entrez Gene ID
55971
Gene Name
BAI1 associated protein 2 like 1
Gene Aliases
IRTKS
Chromosome Location
Chr.7: 98291650 - 98401115 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018842.4 NP_061330.2 8 - 9 1067 81
GenBank mRNA AF119666.2 - 8 - 9 1020 81
AK027109.1 - 4 - 5 522 81
AK309936.1 - 8 - 9 1042 81
BC013888.2 - 8 - 9 1020 81

View Full Product Details