The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015417.4 | NP_056232.2 | 1 - 2 | 277 | 103 |
XM_005260683.4 | XP_005260740.1 | 1 - 2 | 288 | 103 | |
GenBank mRNA | AK313766.1 | - | 1 - 2 | 290 | 103 |
AL080154.1 | - | 1 - 2 | 259 | 103 | |
BC022476.1 | - | 1 - 2 | 236 | 103 | |
BI520334.1 | - | 1 - 2 | 276 | 103 | |
DB030684.1 | - | 1 - 2 | 290 | 103 | |
DB033051.1 | - | 1 - 2 | 270 | 103 | |
EF560745.1 | - | 1 - 2 | 275 | 103 |