The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015417.4 | NP_056232.2 | 2 - 3 | 382 | 77 |
XM_005260683.4 | XP_005260740.1 | 2 - 3 | 393 | 77 | |
GenBank mRNA | AK313766.1 | - | 2 - 3 | 395 | 77 |
AL080154.1 | - | 2 - 3 | 364 | 77 | |
BC022476.1 | - | 2 - 3 | 341 | 77 | |
BI520334.1 | - | 2 - 3 | 381 | 77 | |
DB030684.1 | - | 2 - 3 | 395 | 77 | |
DB033051.1 | - | 2 - 3 | 375 | 77 | |
EF560745.1 | - | 2 - 3 | 380 | 77 |