The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278622.1 | NP_001265551.1 | 9 - 10 | 1364 | 139 |
NM_002873.1 | NP_002864.1 | 8 - 9 | 1469 | 139 | |
NM_133338.2 | NP_579916.1 | 10 - 11 | 1365 | 139 | |
NM_133339.2 | NP_579917.1 | 7 - 8 | 922 | 139 | |
NM_133340.2 | NP_579918.1 | 7 - 8 | 955 | 139 | |
NM_133341.2 | NP_579919.1 | 7 - 8 | 881 | 139 | |
NM_133342.2 | NP_579920.1 | 9 - 10 | 1342 | 139 | |
NM_133343.1 | NP_579921.1 | 9 - 10 | 1307 | 139 | |
NM_133344.2 | NP_579922.1 | 8 - 9 | 1124 | 139 | |
XM_017009679.1 | XP_016865168.1 | 9 - 10 | 1184 | 139 | |
XM_017009680.1 | XP_016865169.1 | 8 - 9 | 1238 | 139 | |
XM_017009681.1 | XP_016865170.1 | 8 - 9 | 1096 | 139 | |
GenBank mRNA | AF017748.1 | - | 8 - 9 | 1094 | 139 |
AF076838.1 | - | 8 - 9 | 1469 | 139 | |
AF098533.1 | - | 7 - 8 | 901 | 139 | |
AF098534.1 | - | 7 - 8 | 827 | 139 | |
AF112263.1 | - | 7 - 8 | 946 | 139 | |
AF126424.1 | - | 10 - 11 | 1364 | 139 | |
AJ001642.1 | - | 8 - 9 | 1133 | 139 | |
AJ004977.1 | - | 8 - 9 | 1129 | 139 | |
AK292487.1 | - | 9 - 10 | 1342 | 139 | |
AL122068.1 | - | 9 - 10 | 1307 | 139 | |
BC032304.1 | - | 7 - 8 | 935 | 139 | |
BX537441.1 | - | 9 - 10 | 1364 | 139 |