The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133338.2 | NP_579916.1 | 3 - 4 | 359 | 159 |
XM_017009679.1 | XP_016865168.1 | 2 - 3 | 178 | 159 | |
GenBank mRNA | AF126424.1 | - | 3 - 4 | 358 | 159 |