The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278622.1 | NP_001265551.1 | 7 - 8 | 1176 | 160 |
NM_002873.1 | NP_002864.1 | 6 - 7 | 1281 | 160 | |
NM_133338.2 | NP_579916.1 | 8 - 9 | 1177 | 160 | |
NM_133339.2 | NP_579917.1 | 5 - 6 | 734 | 160 | |
NM_133340.2 | NP_579918.1 | 5 - 6 | 767 | 160 | |
NM_133341.2 | NP_579919.1 | 5 - 6 | 693 | 160 | |
NM_133342.2 | NP_579920.1 | 7 - 8 | 1154 | 160 | |
NM_133343.1 | NP_579921.1 | 7 - 8 | 1119 | 160 | |
NM_133344.2 | NP_579922.1 | 6 - 7 | 936 | 160 | |
XM_017009679.1 | XP_016865168.1 | 7 - 8 | 996 | 160 | |
XM_017009680.1 | XP_016865169.1 | 6 - 7 | 1050 | 160 | |
XM_017009681.1 | XP_016865170.1 | 6 - 7 | 908 | 160 | |
GenBank mRNA | AF017748.1 | - | 6 - 7 | 906 | 160 |
AF076838.1 | - | 6 - 7 | 1281 | 160 | |
AF085736.2 | - | 5 - 6 | 734 | 160 | |
AF098533.1 | - | 5 - 6 | 713 | 160 | |
AF098534.1 | - | 5 - 6 | 639 | 160 | |
AF112263.1 | - | 5 - 6 | 758 | 160 | |
AF126424.1 | - | 8 - 9 | 1176 | 160 | |
AJ001642.1 | - | 6 - 7 | 945 | 160 | |
AJ004977.1 | - | 6 - 7 | 941 | 160 | |
AK292487.1 | - | 7 - 8 | 1154 | 160 | |
AL122068.1 | - | 7 - 8 | 1119 | 160 | |
BC032304.1 | - | 5 - 6 | 747 | 160 | |
BX537441.1 | - | 7 - 8 | 1176 | 160 |