The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018072.5 | NP_060542.4 | 9 - 10 | 1339 | 171 |
GenBank mRNA | AK095692.1 | - | 9 - 10 | 1302 | 171 |
BC150614.1 | - | 8 - 9 | 1209 | 171 | |
BU102172.1 | - | 4 - 5 | 480 | 171 |