Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL3
Entrez Gene ID
94120
Gene Name
synaptotagmin like 3
Gene Aliases
SLP3
Chromosome Location
Chr.6: 158644881 - 158766298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009991.3 NP_001009991.2 10 - 11 1032 90
NM_001242384.1 NP_001229313.1 12 - 13 1252 90
NM_001242394.1 NP_001229323.1 11 - 12 1644 90
NM_001242395.1 NP_001229324.1 9 - 10 1440 90
NM_001318745.1 NP_001305674.1 8 - 9 966 90
XM_005267215.4 XP_005267272.1 11 - 12 1742 90
XM_005267218.4 XP_005267275.1 11 - 12 1053 90
XM_005267222.4 XP_005267279.1 3 - 4 324 90
XM_006715605.3 XP_006715668.1 12 - 13 1834 90
XM_006715606.3 XP_006715669.1 12 - 13 1755 90
XM_006715611.3 XP_006715674.1 11 - 12 1357 90
XM_011536254.2 XP_011534556.1 12 - 13 1523 90
XM_011536255.2 XP_011534557.1 2 - 3 579 90
XM_017011495.1 XP_016866984.1 9 - 10 1711 90
XM_017011496.1 XP_016866985.1 11 - 12 1923 90
XM_017011497.1 XP_016866986.1 12 - 13 1260 90
XM_017011498.1 XP_016866987.1 8 - 9 1073 90
GenBank mRNA AK055750.1 - 8 - 9 966 90
AK131092.1 - 7 - 8 3395 90
AK298712.1 - 12 - 13 1212 90
BC085612.1 - 9 - 10 978 90
BC100830.1 - 10 - 11 1033 90
BC100831.1 - 10 - 11 1033 90

View Full Product Details