Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL3
Entrez Gene ID
94120
Gene Name
synaptotagmin like 3
Gene Aliases
SLP3
Chromosome Location
Chr.6: 158644881 - 158766298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009991.3 NP_001009991.2 14 - 15 1588 75
NM_001242384.1 NP_001229313.1 16 - 17 1808 75
NM_001242394.1 NP_001229323.1 15 - 16 2200 75
NM_001242395.1 NP_001229324.1 13 - 14 1996 75
NM_001318745.1 NP_001305674.1 12 - 13 1522 75
XM_005267215.4 XP_005267272.1 15 - 16 2298 75
XM_005267218.4 XP_005267275.1 15 - 16 1609 75
XM_005267222.4 XP_005267279.1 7 - 8 880 75
XM_006715605.3 XP_006715668.1 16 - 17 2390 75
XM_006715606.3 XP_006715669.1 16 - 17 2311 75
XM_006715611.3 XP_006715674.1 15 - 16 1913 75
XM_011536254.2 XP_011534556.1 16 - 17 2079 75
XM_011536255.2 XP_011534557.1 6 - 7 1135 75
XM_017011495.1 XP_016866984.1 13 - 14 2267 75
XM_017011496.1 XP_016866985.1 15 - 16 2479 75
XM_017011497.1 XP_016866986.1 16 - 17 1816 75
XM_017011498.1 XP_016866987.1 12 - 13 1629 75
GenBank mRNA AK055750.1 - 12 - 13 1521 75
AK131092.1 - 11 - 12 3951 75
AK298712.1 - 16 - 17 1768 75
AK304191.1 - 7 - 8 1065 75
AL567177.3 - 3 - 4 75
BC085612.1 - 13 - 14 1534 75
BC100830.1 - 14 - 15 1589 75
BC100831.1 - 14 - 15 1589 75

View Full Product Details