Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SETD6
Entrez Gene ID
79918
Gene Name
SET domain containing 6
Gene Aliases
-
Chromosome Location
Chr.16: 58515479 - 58521961 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024860.3 NP_079136.2 2 - 3 175 112
NR_134583.1 2 - 3 175 112
GenBank mRNA AK024801.1 - 2 - 3 131 112
AK297288.1 - 1 - 2 350 112
AK304822.1 - 2 - 3 154 112
BC110433.1 - 2 - 3 159 112
BP338820.1 - 2 - 3 174 112

View Full Product Details