The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145725.1 | NP_001139197.1 | 3 - 4 | 266 | 73 |
NM_017816.2 | NP_060286.1 | 3 - 4 | 364 | 73 | |
XM_011513505.1 | XP_011511807.1 | 3 - 4 | 666 | 73 | |
XM_011513506.2 | XP_011511808.1 | 2 - 3 | 297 | 73 | |
GenBank mRNA | AK000432.1 | - | 3 - 4 | 311 | 73 |
AK290750.1 | - | 3 - 4 | 341 | 73 | |
AL136750.1 | - | 3 - 4 | 266 | 73 | |
BC015796.2 | - | 3 - 4 | 326 | 73 | |
CR533548.1 | - | 1 - 2 | 123 | 73 |