The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018072.5 | NP_060542.4 | 11 - 12 | 1576 | 96 |
GenBank mRNA | AK095692.1 | - | 11 - 12 | 1539 | 96 |
BC150614.1 | - | 10 - 11 | 1446 | 96 |