The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190274.1 | NP_001177203.1 | 9 - 10 | 1228 | 110 |
NM_025133.4 | NP_079409.3 | 9 - 10 | 1017 | 110 | |
XM_005264572.4 | XP_005264629.1 | 9 - 10 | 1222 | 110 | |
XM_005264573.4 | XP_005264630.1 | 9 - 10 | 1175 | 110 | |
XM_017005015.1 | XP_016860504.1 | 9 - 10 | 1175 | 110 | |
XM_017005016.1 | XP_016860505.1 | 9 - 10 | 1041 | 110 | |
XM_017005017.1 | XP_016860506.1 | 9 - 10 | 1041 | 110 | |
GenBank mRNA | AF351618.1 | - | 9 - 10 | 1030 | 110 |
AK022735.1 | - | 5 - 6 | 530 | 110 | |
AK223592.1 | - | 9 - 10 | 1112 | 110 | |
AK292877.1 | - | 9 - 10 | 1067 | 110 | |
AK297444.1 | - | 9 - 10 | 1202 | 110 | |
AK304140.1 | - | 9 - 10 | 932 | 110 | |
AK315108.1 | - | 9 - 10 | 1017 | 110 | |
AY827075.1 | - | 9 - 10 | 1155 | 110 | |
BC043258.1 | - | 9 - 10 | 1119 | 110 | |
BC130445.1 | - | 9 - 10 | 953 | 110 | |
BC136480.1 | - | 9 - 10 | 953 | 110 |