The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166237.1 | NP_001159709.1 | 9 - 10 | 1441 | 66 |
NM_024736.6 | NP_079012.3 | 6 - 7 | 887 | 66 | |
XM_011517301.1 | XP_011515603.1 | 5 - 6 | 1489 | 66 | |
GenBank mRNA | AF318341.1 | - | 7 - 8 | 925 | 66 |
AK022212.1 | - | 6 - 7 | 846 | 66 | |
AK093858.1 | - | 3 - 4 | 891 | 66 | |
AK096216.1 | - | 6 - 7 | 634 | 66 | |
AK123985.1 | - | 4 - 5 | 1335 | 66 | |
AK127941.1 | - | 2 - 3 | 2412 | 66 | |
AK291817.1 | - | 9 - 10 | 1441 | 66 | |
AY008304.1 | - | 6 - 7 | 876 | 66 | |
BC008904.2 | - | 6 - 7 | 872 | 66 | |
BC069000.1 | - | 6 - 7 | 827 | 66 |