The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002767.3 | NP_002758.1 | 2 - 3 | 291 | 148 |
XM_017024866.1 | XP_016880355.1 | 3 - 4 | 298 | 148 | |
XM_017024869.1 | XP_016880358.1 | 2 - 3 | 263 | 148 | |
XM_017024871.1 | XP_016880360.1 | 2 - 3 | 241 | 148 | |
XM_017024875.1 | XP_016880364.1 | 2 - 3 | 171 | 148 | |
GenBank mRNA | AK313087.1 | - | 2 - 3 | 228 | 148 |
AL535304.3 | - | 2 - 3 | 188 | 148 | |
BC106050.1 | - | 2 - 3 | 207 | 148 | |
DC424578.1 | - | 2 - 3 | 291 | 148 |