The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286496.1 | NP_001273425.1 | 6 - 7 | 1204 | 80 |
NM_001286497.1 | NP_001273426.1 | 6 - 7 | 1180 | 80 | |
NM_001286499.1 | NP_001273428.1 | 6 - 7 | 1204 | 80 | |
NM_025049.3 | NP_079325.2 | 6 - 7 | 1180 | 80 | |
XM_011522083.2 | XP_011520385.1 | 6 - 7 | 1296 | 80 | |
XM_011522084.2 | XP_011520386.1 | 6 - 7 | 1128 | 80 | |
GenBank mRNA | AB185926.1 | - | 6 - 7 | 1180 | 80 |
AB185927.1 | - | 6 - 7 | 1180 | 80 | |
AF108138.1 | - | 2 - 3 | 284 | 80 | |
AY498716.1 | - | 5 - 6 | 1104 | 80 | |
BC137503.1 | - | 6 - 7 | 1118 | 80 | |
BC137504.1 | - | 6 - 7 | 1118 | 80 | |
DQ437529.1 | - | 5 - 6 | 1085 | 80 | |
EU084033.1 | - | 6 - 7 | 1175 | 80 |