The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181537.3 | NP_853515.2 | 2 - 3 | 569 | 70 |
GenBank mRNA | AJ564206.1 | - | 2 - 3 | 569 | 70 |