The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 22 - 23 | 2331 | 64 |
NM_018482.3 | NP_060952.2 | 21 - 22 | 2229 | 64 | |
XM_005250925.1 | XP_005250982.1 | 22 - 23 | 2238 | 64 | |
XM_006716563.3 | XP_006716626.1 | 22 - 23 | 2057 | 64 | |
XM_006716564.1 | XP_006716627.1 | 23 - 24 | 2216 | 64 | |
XM_006716565.3 | XP_006716628.1 | 21 - 22 | 2053 | 64 | |
XM_006716566.1 | XP_006716629.1 | 22 - 23 | 2238 | 64 | |
XM_011517052.2 | XP_011515354.1 | 22 - 23 | 2054 | 64 | |
XM_011517053.1 | XP_011515355.1 | 22 - 23 | 2260 | 64 | |
XM_017013467.1 | XP_016868956.1 | 20 - 21 | 2075 | 64 | |
XM_017013468.1 | XP_016868957.1 | 21 - 22 | 2299 | 64 | |
XM_017013469.1 | XP_016868958.1 | 21 - 22 | 2229 | 64 | |
GenBank mRNA | AB033075.1 | - | 15 - 16 | 1462 | 64 |
BC137135.1 | - | 20 - 21 | 2026 | 64 | |
BC144493.1 | - | 21 - 22 | 2128 | 64 | |
BX537768.1 | - | 14 - 15 | 1482 | 64 |