Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEPT7
Entrez Gene ID
989
Gene Name
septin 7
Gene Aliases
CDC10, CDC3, NBLA02942, SEPT7A
Chromosome Location
Chr.7: 35800986 - 35915763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK298453.1 - 2 - 3 125 78
BI669760.1 - 2 - 3 269 78

View Full Product Details