Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2F7
Entrez Gene ID
144455
Gene Name
E2F transcription factor 7
Gene Aliases
-
Chromosome Location
Chr.12: 77021246 - 77065580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203394.2 NP_976328.2 3 - 4 606 79
XM_011537966.2 XP_011536268.1 3 - 4 606 79
GenBank mRNA AK097677.1 - 1 - 2 199 79
BC136366.1 - 3 - 4 445 79
BC136367.1 - 3 - 4 445 79

View Full Product Details