The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003245.3 | NP_003236.3 | 10 - 11 | 1741 | 108 |
GenBank mRNA | AK290324.1 | - | 10 - 11 | 1705 | 108 |
AK291351.1 | - | 10 - 11 | 1705 | 108 | |
AK315236.1 | - | 10 - 11 | 1705 | 108 | |
BC109075.1 | - | 10 - 11 | 1667 | 108 | |
BC109076.1 | - | 10 - 11 | 1667 | 108 | |
BG675531.1 | - | 1 - 2 | 241 | 108 | |
BX284087.1 | - | 1 - 2 | 241 | 108 | |
L10386.1 | - | 10 - 11 | 1685 | 108 |