The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010854.1 | NP_001010854.1 | 12 - 13 | 1584 | 92 |
NM_001320421.1 | NP_001307350.1 | 12 - 13 | 1580 | 92 | |
XM_005267367.1 | XP_005267424.1 | 8 - 9 | 895 | 92 | |
XM_011536497.1 | XP_011534799.1 | 12 - 13 | 1642 | 92 | |
XM_017021044.1 | XP_016876533.1 | 12 - 13 | 1548 | 92 | |
XM_017021045.1 | XP_016876534.1 | 12 - 13 | 1548 | 92 | |
XM_017021046.1 | XP_016876535.1 | 12 - 13 | 1642 | 92 | |
XM_017021047.1 | XP_016876536.1 | 11 - 12 | 1259 | 92 | |
GenBank mRNA | AK126571.1 | - | 3 - 4 | 317 | 92 |
AL832848.1 | - | 11 - 12 | 1262 | 92 | |
BC035865.2 | - | 6 - 7 | 685 | 92 | |
BC048270.2 | - | 6 - 7 | 685 | 92 | |
BX248275.1 | - | 8 - 9 | 876 | 92 |