The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010854.1 | NP_001010854.1 | 18 - 19 | 2233 | 60 |
NM_001320421.1 | NP_001307350.1 | 19 - 20 | 2280 | 60 | |
XM_005267367.1 | XP_005267424.1 | 16 - 17 | 1757 | 60 | |
XM_011536497.1 | XP_011534799.1 | 20 - 21 | 2504 | 60 | |
XM_017021044.1 | XP_016876533.1 | 20 - 21 | 2410 | 60 | |
XM_017021045.1 | XP_016876534.1 | 19 - 20 | 2248 | 60 | |
XM_017021046.1 | XP_016876535.1 | 18 - 19 | 2291 | 60 | |
XM_017021047.1 | XP_016876536.1 | 17 - 18 | 1908 | 60 | |
GenBank mRNA | AK126571.1 | - | 9 - 10 | 966 | 60 |
AL832848.1 | - | 17 - 18 | 1911 | 60 | |
BC035865.2 | - | 13 - 14 | 1529 | 60 | |
BC048270.2 | - | 12 - 13 | 1334 | 60 | |
BX247966.1 | - | 5 - 6 | 572 | 60 | |
BX248275.1 | - | 14 - 15 | 1525 | 60 |