The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004621.5 | NP_004612.2 | 6 - 7 | 2171 | 110 |
XM_011542968.2 | XP_011541270.1 | 6 - 7 | 1778 | 110 | |
XM_017018221.1 | XP_016873710.1 | 4 - 5 | 1913 | 110 | |
XM_017018222.1 | XP_016873711.1 | 6 - 7 | 1673 | 110 | |
GenBank mRNA | AF080394.2 | - | 6 - 7 | 1763 | 110 |
AJ006276.1 | - | 6 - 7 | 2173 | 110 | |
AJ007018.1 | - | 2 - 3 | 110 | ||
AJ271066.1 | - | 6 - 7 | 1746 | 110 | |
AJ271067.1 | - | 4 - 5 | 1398 | 110 | |
AJ271068.1 | - | 5 - 6 | 1581 | 110 | |
AK027769.1 | - | 5 - 6 | 816 | 110 | |
BC093658.1 | - | 6 - 7 | 1810 | 110 | |
BC093660.1 | - | 6 - 7 | 1810 | 110 |