The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006762.2 | NP_006753.1 | 4 - 5 | 482 | 73 |
XM_011542098.2 | XP_011540400.1 | 2 - 3 | 315 | 73 | |
GenBank mRNA | AK292009.1 | - | 4 - 5 | 432 | 73 |
BC106896.1 | - | 4 - 5 | 441 | 73 | |
BC106897.1 | - | 4 - 5 | 441 | 73 | |
D42042.1 | - | 4 - 5 | 412 | 73 | |
U30498.1 | - | 4 - 5 | 413 | 73 | |
U51240.1 | - | 4 - 5 | 466 | 73 |